Anti-ADRB1 Goat Polyclonal Antibody

PRSI45-211EA 420 EUR
PRSI45-211
Anti-ADRB1 Goat Polyclonal Antibody
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 55 kDa band observed in human heart lysates (calculated MW of 51.2 kDa according to NP_000675.1). Recommended concentration: 0.3-1 μg/mL.

ADRB1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ADRB1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ADRB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR