Anti-ADRA2A Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-210EA 420 EUR
PRSI45-210
Anti-ADRA2A Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Preliminary experiments gave a band at approximately 37 kDa and a minor band at approximately 23 kDa in human brain lysates after 1 μg/mL antibody staining. Both detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional bands). ADRA2A antibody has been used in mouse ICC/IF assays.

ADRA2A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ADRA2A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ADRA2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR