Anti-ADH7 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-208EA 345 EUR
PRSI45-208
Anti-ADH7 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 26 kDa band in human duodenum lysate and liver and of rat stomach after 0.2 μg/mL antibody staining.5 kDa according to NP_000664.2. The 26 kDa band was successfully blocked by incubation with the immunizing peptide.

ADH7 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ADH7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ADH7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR