Anti-ADORA2B Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-206EA 420 EUR
PRSI45-206
Anti-ADORA2B Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Immunohistochemistry: In paraffin embedded human kidney shows cytoplasm staining in cells of renal tubules. Recommended concentration, 3 μg/mL.

Adenosine A2 beta Receptor antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Adenosine A2 beta Receptor antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ADORA2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR