Anti-ADAM33 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-204EA 420 EUR
PRSI45-204
Anti-ADAM33 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: experiments gave bands at approximately 80-90 kDa and strong ladder from 26 to 38 kDa in human prostate lysates after 0.3 μg/mL antibody staining. The observation of multiple bands and the 38 kDa band correspond to earlier findings in literature with different antibodies (Powell et al, Am J Respir Cell Mol Biol. 2004 Jul;31 (1) :13-21; PMID: 14742294). This protein has a calculated MW of 87.7 kDa according to NP_079496.1. Recommended concentration: 0.1-0.3 μg/mL.

ADAM33 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ADAM33 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ADAM33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR