Anti-ACYP1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-203EA 553 EUR
PRSI45-203
Anti-ACYP1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 15 kDa band observed in human testis lysates (calculated MW of 11.3 kDa according to NP_001098.1). Recommended concentration: 0.01-0.03 μg/mL.

ACYP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ACYP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ACYP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR