Anti-ACHE Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-193EA 420 EUR
PRSI45-193
Anti-ACHE Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Approximately 70 kDa band observed in human cerebellum lysates (calculated MW of 67.8 kDa according to NP_000656.1). Recommended concentration: 1-3 μg/mL.

ACHE antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ACHE antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ACHE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR