Anti-ACAP1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-191EA 553 EUR
PRSI45-191
Anti-ACAP1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution >1:32,000. Western Blot: Approximately 80-90 kDa band observed in Jurkat lysates (predicted MW of 85 kDa according to NP_055531). Recommended for use at 0.5-2 μg/mL.

ACAP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ACAP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ACAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR