Anti-ACACB Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-189EA 553 EUR
PRSI45-189
Anti-ACACB Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 28 kDa band in human heart lysates after 0.5 μg/mL antibody staining.2. The 28 kDa band was successfully blocked by incubation with the immunizing peptide.

ACACB antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ACACB antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ACACB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR