Anti-ABHD4 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-184EA 420 EUR
PRSI45-184
Anti-ABHD4 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 48 kDa band in human brain (cerebellum, cerebral cortex and frontal cortex) lysates after 1 μg/mL antibody staining. The 48 kDa band was successfully blocked by incubation with the immunizing peptide.

ABHD4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ABHD4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ABHD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR