Anti-ABCC4 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-181EA 420 EUR
PRSI45-181
Anti-ABCC4 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 170 kDa band observed in rat renal cortex membranes (calculated MW of 149 kDa according to rat NP_596902.1 and 150 kDa according to human NP_005836.2). Recommended concentration: 1-3 μg/mL. Immunohistochemistry: In paraffin embedded human prostate shows strong cytoplasmic staining in small flat basal cells and weak cytoplasmic staining in the columnar secretory cells of the epithelium. Recommended concentration, 2-4 μg/mL.

ABCC4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ABCC4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ABCC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR