Anti-TRIM5 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-163EA 420 EUR
PRSI45-163
Anti-TRIM5 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 37 kDa band observed in human brain lysates (calculated MW of 37.7 kDa according to NP_149084.1, isoform delta). an additional faint 40 kDa band consistent with calculated MW of 40.1 kDa according to NP_149083.1 (isoform gamma) is also observed. Recommended concentration: 1-3 μg/mL. Immunohistochemistry: In paraffin embedded human tonsil, stains a group of cells in the interfollicular area, below the epithelium of tonsillar criptae and weak in germinal centre. Recommended concentration, 1.0 μg/mL.

TRIM5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TRIM5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRIM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR