Anti-SPP1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-152EA 553 EUR
PRSI45-152
Anti-SPP1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 30 kDa band observed in lysates of cell lines Jurkat and MOLT4. This band corresponds to the cleavage product from the 60 kDa glycoprotein (calculated MW of 33.8 kDa according to NP_000573.1). Recommended concentration: 1-3 μg/mL.

Osteopontin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Osteopontin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SPP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR