Anti-SLC18A2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-146EA 420 EUR
PRSI45-146
Anti-SLC18A2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: A faint band of appr. 50 kDa band observed in human placenta lysates (calculated MW of 55.7 kDa according to NP_003045). Recommended concentration: 0.3-1.0 μg/mL.

VMAT2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

VMAT2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLC18A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR