Anti-SERPINE1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-142EA 420 EUR
PRSI45-142
Anti-SERPINE1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 45 kDa band observed in human duodenum, liver and lung lysates (calculated MW of 45.1 kDa according to NP_000593.1). Recommended concentration: 0.1-0.3 μg/mL. Immunohistochemistry: In paraffin embedded human placenta shows strong staining of cytotrophoblasts. Recommended concentration, 4-6 μg/mL. Also been successfully stained paraffin embedded human breast carcinoma tissue. Enzyme Immunoassay: Sandwich-type ELISA with increasing amount of recombinant PAI1 captured by a rabbit antibody. Recommended reporter concentration: 1.5-3 μg/mL.

PAI-1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PAI-1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SERPINE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR