Anti-SEC61A1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-141EA 420 EUR
PRSI45-141
Anti-SEC61A1 Goat Polyclonal Antibody
Antikörper
Western Blot: Approximately 40 kDa band seen in Daudi cell lysate. Recommended for use at 2-4 μg/mL.

Sec61 alpha antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Sec61 alpH a antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SEC61A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR