Anti-RBP1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-136EA 420 EUR
PRSI45-136
Anti-RBP1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 16 kDa band observed in Murine NIH 3T3 lysates (calculated MW of 15.9 kDa according to NP_002890.1). Recommended concentration: 0.1-0.3 μg/mL. Immunohistochemistry: In paraffin embedded human pancreas shows vesicular staining in the acinar cytoplasm but oriented towards the interlobular space. Recommended concentration, 5-10 μg/mL.

RBP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

RBP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR