Anti-RANGAP1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-135EA 420 EUR
PRSI45-135
Anti-RANGAP1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 70 kDa band observed in human brain and 3T3 lysates (predicted MW of 68 kDa according to NP_002874). Recommended for use at 0.1-0.5 μg/mL. Immunohistochemistry: In paraffin embedded human liver shows vesicular staining in the cytoplasm of hepatocytes. Recommended concentration, 2-4 μg/mL.

RANGAP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

RANGAP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RANGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR