Anti-PPP2CA / PPP2CB Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-119EA 420 EUR
PRSI45-119
Anti-PPP2CA / PPP2CB Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 35 kDa band observed in human cerebellum, mouse brain and rat brain lysates (calculated MW of 35.6 kDa according to NP_002706.1and NP_004147.1). Recommended concentration: 0.05-0.2 μg/mL.

PPP2C antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PPP2C antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PPP2CA / PPP2CB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR