Anti-PDHX Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-107EA 420 EUR
PRSI45-107
Anti-PDHX Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 50 kDa band observed in human pancreas lysate (calculated MW of 54.2 kDa according to NP_003468.1). In transfected HEK293 transiently expressing PDHX a band of approximately 50 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.1-1 μg/mL. Immunohistochemistry: In paraffin embedded human pancreas, shows cytoplasmic staining of acinar cells, islets of Langerhans and peri-pancreatic fat cells. Recommended concentration, 1-3 μg/mL.

Pyruvate Dehydrogenase antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Pyruvate dehydrogenase antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PDHX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR