Anti-PACSIN1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-103EA 553 EUR
PRSI45-103
Anti-PACSIN1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: experiments gave bands at approximately 50 kDa and 37 kDa in human brain lysate after 1 μg/mL antibody staining. Calculated MW is 50 kDa according to NP_065855.1. Both bands were successfully blocked by incubation with the immunizing peptide. Recommended concentration: 1-3 μg/mL.

PACSIN1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PACSIN1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PACSIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR