Anti-MYD88 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-095EA 553 EUR
PRSI45-095
Anti-MYD88 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 33 kDa band observed in humanthymus lysates (calculated MW of 33.2 kDa according to NP_002459.1). Recommended concentration: 0.03-0.1 μg/mL. Immunohistochemistry: In paraffin embedded human tonsil shows staining of clusters of cells some of which are likely plasma cells. Recommended concentration, 4-6μg/ml

MYD88 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MYD88 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MYD88
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR