Anti-MELK Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-089EA 420 EUR
PRSI45-089
Anti-MELK Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: experiments gave bands at approximately 75 kDa and 150 kDa in human testis lysates and in cell lines HeLa and Jurkat after 0.1 μg/mL antibody staining. The150 kDa band is explained by glycosylation of this member of the AMPK family. This protein has a calculated MW of 74.6 kDa according to NP_055606.1. Recommended concentration: 0.1-0.3 μg/mL.

MELK antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MELK antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MELK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR