Anti-IRF8 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-073EA 553 EUR
PRSI45-073
Anti-IRF8 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 45-50 kDa band seen in K562 lysate (Predicted MW of 47 kDa according to NP_002154). Recommended for use at 0.5-2 μg/mL.

ICSBP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ICSBP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IRF8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR