Anti-HSP90AB1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-071EA 420 EUR
PRSI45-071
Anti-HSP90AB1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 100 kDa band observed in NIH/3T3 lysate (calculated MW of 92.5 kDa according to NP_003290.1). Recommended concentration: 0.01-0.03 μg/mL. Immunohistochemistry: In paraffin embedded human liver shows staining in some hepatocyte nuclei and weak cytoplasm staining of hepatocytes. Recommended concentration, 3-5 μg/mL.

HSP90B1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

HSP90B1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HSP90AB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR