Anti-GPX1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-065EA 420 EUR
PRSI45-065
Anti-GPX1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 22 kDa band observed in human liver and kidney lysates (calculated MW of 22.1 kDa according to NP_000572.2). Recommended concentration: 1-3 μg/mL. Immunohistochemistry: In paraffin embedded human cerebral cortex shows staining of microglia. Recommended concentration, 3-5 μg/mL.

Glutathione Peroxidase 1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Glutathione peroxidase 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR