Anti-GIPC1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-064EA 420 EUR
PRSI45-064
Anti-GIPC1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 38 kDa and strong 23-24 kDa bands observed in HeLa lysate, Approximately 24 kDa also observed in human pancreas, kidney and muscle lysates (calculated MW of 36.0 kDa according to NP_005707.1, NP_974197.1 and NP_974199.1 and 26.1 kDa according to NP_974198.1 and NP_974223.1). Recommended concentration: 0.03-0.1 μg/mL.

GIPC1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GIPC1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GIPC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR