Anti-FTCD Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-057EA 420 EUR
PRSI45-057
Anti-FTCD Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 60 kDa band observed in human liver lysates (calculated MW of 58.9 kDa according to NP_006648.1and NP_996848.1). Recommended concentration: 0.01-0.03 μg/mL.

FTCD antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FTCD antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FTCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR