Anti-CTTN Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-044EA 420 EUR
PRSI45-044
Anti-CTTN Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 80 kDa band observed in mouse and rat brain lysates (calculated MW of 61.6 kDa according to human NP_005222.2 and of 61.2 kDa according to mouse NP_031829.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Paradis et al, J Cell Sci. 2008 Jun 15;121 (Pt 12) :1965-72; PMID 18495841). Recommended concentration: 0.01-0.03 μg/mL.

Cortactin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Cortactin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CTTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR