Anti-CST3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-043EA 420 EUR
PRSI45-043
Anti-CST3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 18 kDa band observed in mouse Tesits and mouse kidney lysates (calculated MW of 15.5 kDa according to NP_034106.2). Recommended concentration: 0.01-0.03 μg/mL.

Cystatin C antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Cystatin C antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CST3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR