Anti-CHGA Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-039EA 420 EUR
PRSI45-039
Anti-CHGA Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 50 kDa band observed in human kidney lysates (calculated MW of 50.7 kDa according to NP_001266.1). Recommended concentration: 0.1-0.3 μg/mL.

Chromogranin A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Chromogranin A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CHGA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR