Anti-CHAT Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-038EA 420 EUR
PRSI45-038
Anti-CHAT Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 70 kDa band observed in human placenta lysates (calculated MW of 70.5 kDa according to NP_065574 and NP_066264). Recommended concentration: 0.03-0.3 μg/mL.

ChAT antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ChAT antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CHAT
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR