Anti-CHAT Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-037EA 553 EUR
PRSI45-037
Anti-CHAT Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 75-80 kDa band observed in mouse and rat brain lysates (calculated MW of 72.3 kDa according to NP_034021.1). Recommended concentration: 0.2-0.5 μg/mL.

ChAT antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ChAT antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CHAT
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR