Anti-CBX3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-029EA 420 EUR
PRSI45-029
Anti-CBX3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 20 kDa band observed in lysates of A431, HeLa and HepG2. (calculated MW of 20.8 kDa according to NP_009207.2 and NP_057671.2). Recommended concentration: 0.3-1.0 μg/mL. Immunohistochemistry: In paraffin embedded human placenta shows strong nuclear staining in trophoblasts. Recommended concentration, 2-3 μg/mL.

CBX3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CBX3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CBX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR