Anti-CAT Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-026EA 553 EUR
PRSI45-026
Anti-CAT Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 60 kDa band observed in human kidney and liver lysates (calculated MW of 59.8 kDa according to NP_001743.1). Recommended concentration: 0.3-1 μg/mL. Immunohistochemistry: Immunohistochemistry: In paraffin embedded human liver shows vesicular staining in the cytoplasm of hepatocytes. Recommended concentration, 2-4 μg/mL.

Catalase antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Catalase antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR