Anti-BIRC7 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-024EA 420 EUR
PRSI45-024
Anti-BIRC7 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: experiments gave bands at approximately 30 kDa and 35 kDa in human acute lymphoblastic leukaemia MOLT4 lysate after 0.3 μg/mL antibody staining. These bands correspond to the reported isoform.

BIRC7 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

BIRC7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BIRC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR