Anti-ARPC2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-011EA 420 EUR
PRSI45-011
Anti-ARPC2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 33-34 kDa band observed in Hela and human skeletal muscle lysates (predicted MW of 34 kDa according to NP_005722). Recommended for use at 0.1-0.5 μg/mL. Immunohistochemistry: In paraffin embedded human spleen shows cytoplasm staining in some splenocytes in and around a T-cell area. Recommended concentration, 3-5 μg/mL.

ARPC2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ARPC2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ARPC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR