Anti-AKT3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-005EA 553 EUR
PRSI45-005
Anti-AKT3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: A 50-55 kDa band observed in human hepatoblastoma HepG2 lysates (calculated MW of 55.8 kDa according to NP_005456 and 54.0 kDa according to NP_859029). Recommended concentration: 1-3 μg

AKT3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

AKT3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: AKT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR