Anti-TUBB3 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI43-094EA 553 EUR
PRSI43-094
Anti-TUBB3 Goat Polyclonal Antibody
Antikörper
The starting concentration recommended for Western Blot is 1-3μg/mL.

TUBB3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TUBB3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TUBB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR