Anti-GEM Goat Polyclonal Antibody

Lieferant: ProSci
PRSI43-052EA 553 EUR
PRSI43-052
Anti-GEM Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. The starting concentration recommended for Western Blot is 1-3μg/mL.

GEM antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GEM antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GEM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR