Anti-HDAC1 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI43-003EA 553 EUR
PRSI43-003
Anti-HDAC1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1000. Western Blot: Approx 55 kDa band observed in lysates of cell line NIH3T3 (calculated MW of 55.1 kDa according to Human NP_032254.1 and Mouse NP_032254.1). Recommended concentration: 0.3-1 μg/mL.

Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Hdac1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HDAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR