Anti-TNFRSF1A Goat Polyclonal Antibody

Lieferant: ProSci
PRSI42-996EA 420 EUR
PRSI42-996
Anti-TNFRSF1A Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 32000. Western Blot: Approx 52 kDa band observed in Human Lung lysates (calculated MW of 50.5 kDa according to NP_001056.1). Recommended concentration: 0.3-1 μg/mL.

Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TNFRSF1A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TNFRSF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR