Anti-SFTPA1 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI42-993EA 553 EUR
PRSI42-993
Anti-SFTPA1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 32000. Western Blot: Approx 30 kDa band observed in Human Lung lysates (calculated MW of 27.7 kDa according to NP_001087239.2). Recommended concentration: 0.01-0.03 μg/mL.

Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

SFTPA1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SFTPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR