Anti-ZAP70 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI42-992EA 420 EUR
PRSI42-992
Anti-ZAP70 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 32000. Western Blot: Approx 70 kDa band observed in lysates of cell lines Jurkat and Molt4 (calculated MW of 69.9 kDa according to NP_001070.2). Recommended concentration: 0.3-1 μg/mL.

Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ZAP70 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ZAP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR