Anti-HMHA1 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI42-988EA 553 EUR
PRSI42-988
Anti-HMHA1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 128000. Western Blot: Preliminary experiments gave an approx 200 kDa band in lysates of cell lines Jurkat and Molt4 after 0.05 μg/mL antibody staining.

Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

HMHA1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HMHA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR