Anti-MAPK9 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI42-985EA 420 EUR
PRSI42-985
Anti-MAPK9 Goat Polyclonal Antibody
Antikörper
Western Blot: Approx 48 kDa band observed in Human cerebellum and Mouse Brain lysates (calculated MW of 48.3 kDa according to NP_620709.1). Recommended concentration: 0.5-1.5 μg/mL.

Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MAPK9 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAPK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR