Microplates, 96 and 384 well, high content imaging with glass bottom, Corning® BioCoat™
Lieferant: Corning Life Sciences
BioCoat™ Corning® BioCoat™
BDAA4582EA 460.08 EUR
BDAA4582 BDAA4583 BDAA4584 BDAA4585 BDAA4586 BDAA4587
Microplates, 96 and 384 well, high content imaging with glass bottom, Corning® BioCoat™
Mikrotiterplatten HTS-Mikrotiterplatten
High optical quality, glass bottom, black microplates are ideal for performing high content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.
  • High optical quality and scratch-resistant glass
  • Glass bottom thickness of 200 µm is well suited for imaging microscopy
  • Bottom flatness <50 µm to ensure planarity for imaging devices
  • Low background fluorescence and minimal cross-talk provides the highest possible optical quality for cell-based assays
  • Half area 96-well microplate reduces reagent consumption