San Joaquin Soil, trace elements, NIST-2709A standard
Lieferant: LGC Standards
PROONIST-2709AEA 1375 EUR
PROONIST-2709A
San Joaquin Soil, trace elements, NIST-2709A standard
This standard reference material is intended primarily for use in the analysis of soils, sediments and other materials of a similar matrix.
One unit consists of 50 g of dried, powdered, agricultural soil.
Standards: Traceable to NIST and SRM numbers.