Marine sediment - trace elements, NIST-2702
Lieferant: LGC Standards
PROONIST-2702EA 1024 EUR
PROONIST-2702
Marine sediment - trace elements, NIST-2702
This reference standard is intended for use in evaluating analytical methods for the determination of selected elements in marine and fresh water sediment and similar matrices.
A unit consists of a bottle containing 50 g of radiation-sterilised, freeze-dried sediment material.
Standards: Traceable to NIST and SRM.