House dust, standard reference material, NIST-2585
Lieferant: LGC Standards
PROONIST-2585EA 1236 EUR
PROONIST-2585
House dust, standard reference material, NIST-2585
Standards Umweltanalyse-Standards
This reference standard is intended for use in evaluating analytical methods for the determination of selected of selected polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyl (PCB) congers, chlorinated pesticides and polybrominated diphenyl ether (PBDE) cogeners in house dust and similar matrices.
Standards: Traceable to NIST and SRM.